MilliporeSigma
  • Carboxyl-ester lipase maturity-onset diabetes of the young is associated with development of pancreatic cysts and upregulated MAPK signaling in secretin-stimulated duodenal fluid.
Carboxyl-ester lipase maturity-onset diabetes of the young is associated with development of pancreatic cysts and upregulated MAPK signaling in secretin-stimulated duodenal fluid.
Diabetes (2013-09-26)
Helge Ræder, Fiona E McAllister, Erling Tjora, Shweta Bhatt, Ingfrid Haldorsen, Jiang Hu, Stefan M Willems, Mette Vesterhus, Abdelfattah El Ouaamari, Manway Liu, Maria B Ræder, Heike Immervoll, Dag Hoem, Georg Dimcevski, Pål R Njølstad, Anders Molven, Steven P Gygi, Rohit N Kulkarni
ABSTRACT
Carboxyl-ester lipase (CEL) maturity-onset diabetes of the young (MODY) is a monogenic form of diabetes and pancreatic exocrine dysfunction due to mutations in the CEL gene encoding CEL. The pathogenic mechanism for diabetes development is unknown. Since CEL is expressed mainly in pancreatic acinar cells, we asked whether we could find structural pancreatic changes in CEL-MODY subjects during the course of diabetes development. Furthermore, we hypothesized that the diseased pancreas releases proteins that are detectable in pancreatic fluid and potentially reflect activation or inactivation of disease-specific pathways. We therefore investigated nondiabetic and diabetic CEL-mutation carriers by pancreatic imaging studies and secretin-stimulated duodenal juice sampling. The secretin-stimulated duodenal juice was studied using cytokine assays, mass spectrometry (MS) proteomics, and multiplexed MS-based measurement of kinase activities. We identified multiple pancreatic cysts in all eight diabetic mutation carriers but not in any of the four nondiabetic mutation carriers or the six healthy controls. Furthermore, we identified upregulated mitogen-activated protein kinase (MAPK) target proteins and MAPK-driven cytokines and increased MAPK activity in the secretin-stimulated duodenal juice. These findings show that subjects with CEL-MODY develop multiple pancreatic cysts by the time they develop diabetes and that upregulated MAPK signaling in the pancreatic secretome may reflect the pathophysiological development of pancreatic cysts and diabetes.
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g