MilliporeSigma
  • (+)-rumphiin and polyalthurea, new compounds from the stems of Polyalthia rumphii.
(+)-rumphiin and polyalthurea, new compounds from the stems of Polyalthia rumphii.
Natural product communications (2013-12-21)
Tian-Shan Wang, You-Ping Luo, Jing Wang, Meng-Xiong He, Ming-Guo Zhong, Ying Li, Xiao-Ping Song
ABSTRACT
Two new compounds, (+)-rumphiin (3) and polyalthurea (7), together with seven known ones, 3,4,5-trimethoxy benzoic acid (1), (-)-seselinone (2), cannabisin D (4), allantoin (5), oxostephanine (6) and a mixture of beta-sitosterol (8) and stigmasterol (9) were isolated from the stems of Polyalthia rumphii. The chemical structures of 3 and 7 were elucidated by the combination of spectroscopic data, and the absolute configuration of 3 at C-2 was determined by the matrix method to be R. All compounds were evaluated for their cytotoxicity on four human cancer cell lines, which demonstrated that 3 was a moderate bioactive lignan, and 6 showed significant anticancer activity against SPC-A-1 and BEL-7402 cell lines with IC50 values of 1.47 and 1.73 microg/mL, respectively.
Product Description
Supelco
Allantoin, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Allantoin, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Stigmasterol, ~95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Allantoin, ≥98.0% (N)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mg
Estimated to ship on January 20, 2026
from MILWAUKEE
$122.00
10 mg
Estimated to ship on January 27, 2025
from MILWAUKEE
$555.00
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$227.00
25 mg
In Stock
Details...
$425.00
$382.50
100 mg
In Stock
Details...
$1,290.00
$1,161.00
Sigma-Aldrich
β-Sitosterol, ≥70%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$97.60
Sigma-Aldrich
3,4,5-Trimethoxybenzoic acid, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
Estimated to ship on March 21, 2025
$133.00
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
In Stock
Details...
$46.90
$43.62
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$113.00
250 g
In Stock
Details...
$185.00
1 kg
In Stock
Details...
$489.00