MilliporeSigma
  • Starch/polyvinyl alcohol blended materials used as solid carbon source for tertiary denitrification of secondary effluent.
Starch/polyvinyl alcohol blended materials used as solid carbon source for tertiary denitrification of secondary effluent.
Journal of environmental sciences (China) (2014-02-06)
Peng Li, Jiane Zuo, Wei Xing, Lei Tang, Xiangyang Ye, Li Zaixing, Lin Yuan, Kaijun Wang, Hongtao Zhang
ABSTRACT
Starch/polyvinyl alcohol (PVA) blended materials for using as a solid carbon source (SCS) were prepared by blending PVA and gelatinized starch in an aqueous solution system, in which PVA served as framework material and starch as carbon source. The optimization of starch content and temperature effects were investigated. It was indicated that higher denitrification efficiency could be achieved with more starch in the materials. The average specific denitrification rates were 0.93, 0.66, 0.37 and 0.36 mg/(g x day) corresponding to starch content of 70%, 60%, 40% and 30% respectively at 37 degrees C. The denitrification rates increased when operating temperature was raised from 23 degrees C to 30 degrees C and then 37 degrees C. The mechanism of carbon release was analyzed incorporating the experimental results of abiotic release in deionized water. The organic carbon was mainly hydrolyzed by microbes, and the biological release efficiencies were at the range of 89.2% to 96.0%. A long-term experiment with a continuous flow reactor with SCS material containing 70% starch was conducted to gain some experience for practical application. When the influent nitrate concentration was in the range of 35.2 to 39.1 mg/L, hydraulic retention time of 4 hr, and operating temperature of 30 degrees C, a nitrogen removal efficiency up to 94.6% and denitrification rate of 0.217 kg/(m3 x day) was achieved. The starch-based materials developed in this study can be used as a solid carbon source for tertiary nitrogen removal from secondary effluent.
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Starch from potato, Soluble
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Starch from potato, Powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Starch from wheat, Unmodified
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), SAJ first grade, degree of polymerization 500
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod