MilliporeSigma
  • Modification of agarose: 6-aminoagarose mediated syntheses of fluorogenic pyridine carboxylic acid amides.
Modification of agarose: 6-aminoagarose mediated syntheses of fluorogenic pyridine carboxylic acid amides.
Carbohydrate polymers (2014-04-12)
Stalin Kondaveeti, Gaurav K Mehta, A K Siddhanta
ABSTRACT
A facile 6-aminoagarose (AA) mediated synthesis of new fluorogenic amides of agarose with nicotinic (AA-NA) and picolinic acids (AA-PA) employing carbodiimide chemistry have been described. 6-Amino agarose (AA) was synthesized in a facile Mitsunobu-inspired microwave mediated method involving the reaction of agarose with phthalimide in presence of diisopropyl azodicarboxylate and triphenylphosphene (DIAD/TPP) followed by hydrazinolysis. All compounds were characterized by GPC, UV spectrophotometry, fluorescence spectroscopy, FT-IR, (1)H and (13)C NMR spectra. The fluorescence emissions (λmax 430 and 412 nm) of 1 × 10(-3)M solutions of AA-NA and AA-PA in water were significantly higher (ca. 82% and ca. 90%) than those of the molar equivalents (0.2mg) of NA and PA present in the 1 × 10(-3)M solutions of the amides, respectively. These fluorogenic pyridine carboxylic acid amides of agarose may find applications as sensors in biomedical and pharmaceutical industries.
Sigma-Aldrich
Phthalimide, ≥99%