MilliporeSigma
  • Protection against paraoxon toxicity by an intravenous pretreatment with polyethylene-glycol-conjugated recombinant butyrylcholinesterase in macaques.
Protection against paraoxon toxicity by an intravenous pretreatment with polyethylene-glycol-conjugated recombinant butyrylcholinesterase in macaques.
Chemico-biological interactions (2014-01-05)
Yvonne J Rosenberg, Jeffery Gearhart, Lingjun Mao, Xiaoming Jiang, Segundo Hernandez-Abanto
ABSTRACT
Recombinant (r) butyrylcholinesterase (rBChE) produced in CHO cells is being developed as a prophylactic countermeasure against neurotoxicity resulting from exposure to organophosphates (OPs) in the form of pesticides and nerve agents. To evaluate the efficacy of a parenteral pretreatment, a PEGylated macaque (Ma) form of rBChE was administered into homologous animals to ensure good plasma retention without immunogenicity. Thus, macaques were administered PEG-rMaBChE at either 5 or 7mg/kg intravenously (i.v.) and exposed subcutaneously to 12μg/kg of the potent pesticide paraoxon (Px) at 1h or at 1 and 72h, respectively. Protection was measured by the ability of rBChE prophylaxis to prevent the inhibition of circulating acetylcholinesterase on red blood cells (RBC-AChE). In rBChE-pretreated animals, no inhibition of RBC-AChE activity after the first Px exposure and only a 10-20% reduction after the second exposure were observed as compared to a 75% RBC-AChE inhibition usually obtained without pretreatment. In addition, these studies raised other interesting issues. The lipophilic nature of Px, appears to result in early and transient inhibition of RBC-AChE as a result of transfer of OP bound to RBC even in BChE-pretreated animals. The protection by a single injection of rBChE against two administrations of Px represents the first example of protection by an i.v. rBChE pretreatment against a pesticide such as Px and bodes well for a parenteral rHuBChE pretreatment as an OP countermeasure in humans.
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)