MilliporeSigma
  • Results of a multicenter, controlled, randomized clinical trial evaluating the combination of piperacillin/tazobactam and tigecycline in high-risk hematologic patients with cancer with febrile neutropenia.
Results of a multicenter, controlled, randomized clinical trial evaluating the combination of piperacillin/tazobactam and tigecycline in high-risk hematologic patients with cancer with febrile neutropenia.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2014-04-16)
Giampaolo Bucaneve, Alessandra Micozzi, Marco Picardi, Stelvio Ballanti, Nicola Cascavilla, Prassede Salutari, Giorgina Specchia, Rosa Fanci, Mario Luppi, Laura Cudillo, Renato Cantaffa, Giuseppe Milone, Monica Bocchia, Giovanni Martinelli, Massimo Offidani, Anna Chierichini, Francesco Fabbiano, Giovanni Quarta, Valeria Primon, Bruno Martino, Annunziata Manna, Eliana Zuffa, Antonella Ferrari, Giuseppe Gentile, Robin Foà, Albano Del Favero
ABSTRACT
Empiric antibiotic monotherapy is considered the standard of treatment for febrile neutropenic patients with cancer, but this approach may be inadequate because of the increasing prevalence of infections caused by multidrug resistant (MDR) bacteria. In this multicenter, open-label, randomized, superiority trial, adult, febrile, high-risk neutropenic patients (FhrNPs) with hematologic malignancies were randomly assigned to receive piperacillin/tazobactam (4.5 g intravenously every 8 hours) with or without tigecycline (50 mg intravenously every 12 hours; loading dose 100 mg). The primary end point was resolution of febrile episode without modifications of the initial allocated treatment. Three hundred ninety FhrNPs were enrolled (combination/monotherapy, 187/203) and were included in the intention-to-treat analysis (ITTA). The ITTA revealed a successful outcome in 67.9% v 44.3% of patients who had received combination therapy and monotherapy, respectively (127/187 v 90/203; absolute difference in risk (adr), 23.6%; 95% CI, 14% to 33%; P < .001). The combination regimen proved better than monotherapy in bacteremias (adr, 32.8%; 95% CI, 19% to 46%; P < .001) and in clinically documented infections (adr, 36%; 95% CI, 9% to 64%; P < .01). Mortality and number of adverse effects were limited and similar in the two groups. The combination of piperacillin/tazobactam and tigecycline is safe, well tolerated, and more effective than piperacillin/tazobactam alone in febrile, high-risk, neutropenic hematologic patients with cancer. In epidemiologic settings characterized by a high prevalence of infections because of MDR microorganisms, this combination could be considered as one of the first-line empiric antibiotic therapies.
Sigma-Aldrich
Piperacillin sodium salt, penicillin analog
Supelco
Piperacillin sodium salt, analytical standard
Sigma-Aldrich
Minocycline hydrochloride, powder