MilliporeSigma
  • Mode of action: oxalate crystal-induced renal tubule degeneration and glycolic acid-induced dysmorphogenesis--renal and developmental effects of ethylene glycol.
Mode of action: oxalate crystal-induced renal tubule degeneration and glycolic acid-induced dysmorphogenesis--renal and developmental effects of ethylene glycol.
Critical reviews in toxicology (2006-01-19)
Richard A Corley, M E Meek, Edward W Carney
ABSTRACT
Ethylene glycol can cause both renal and developmental toxicity, with metabolism playing a key role in the mode of action (MOA) for each form of toxicity. Renal toxicity is ascribed to the terminal metabolite oxalic acid, which precipitates in the kidney in the form of calcium oxalate crystals and is believed to cause physical damage to the renal tubules. The human relevance of the renal toxicity of ethylene glycol is indicated by the similarity between animals and humans of metabolic pathways, the observation of renal oxalate crystals in toxicity studies in experimental animals and human poisonings, and cases of human kidney and bladder stones related to dietary oxalates and oxalate precursors. High-dose gavage exposures to ethylene glycol also cause axial skeletal defects in rodents (but not rabbits), with the intermediary metabolite, glycolic acid, identified as the causative agent. However, the mechanism by which glycolic acid perturbs development has not been investigated sufficiently to develop a plausible hypothesis of mode of action, nor have any cases of ethylene glycol-induced developmental effects been reported in humans. Given this, and the variations in sensitivity between animal species in response, the relevance to humans of ethylene glycol-induced developmental toxicity in animals is unknown at this time.
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Supelco
Ethylene glycol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Supelco
Glycolic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00
Sigma-Aldrich
Ethylene glycol 5 M solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
Estimated to ship on March 21, 2025
$279.00