MilliporeSigma
  • Administration of erythromycin before endoscopy in upper gastrointestinal bleeding: a meta-analysis of randomized controlled trials.
Administration of erythromycin before endoscopy in upper gastrointestinal bleeding: a meta-analysis of randomized controlled trials.
Saudi journal of gastroenterology : official journal of the Saudi Gastroenterology Association (2013-09-21)
Shoba Theivanayagam, Roxanne G Lim, William J Cobell, Jayashree T Gowda, Michelle L Matteson, Abhishek Choudhary, Matthew L Bechtold
ABSTRACT
Erythromycin infusion before endoscopy in upper gastrointestinal bleeding (UGIB) has been hypothesized to aid in visualization and reduce the need for second-look endoscopy; however, the results have been controversial. To evaluate further, we performed a meta-analysis comparing the efficacy of erythromycin infusion before endoscopy in acute UGIB. Multiple databases were searched (March 2013). Only randomized controlled trials were included in the analysis. A meta-analysis for the effect of erythromycin or no erythromycin before endoscopy in UGIB were analyzed by calculating pooled estimates of primary (visualization of gastric mucosa and need for second endoscopy) and secondary (units of blood transfused, length of hospital stay, duration of the procedure) outcomes. Statistical analysis was performed using RevMan 5.1 (Copenhagen: The Nordic Cochrane Centre, The Cochrane Collaboration). Six studies (N = 558) met the inclusion criteria. Erythromycin infusion before endoscopy in UGIB demonstrated a statistically significant improvement in visualization of the gastric mucosa [odds ratio (OR) 3.43; 95% confidence interval (CI): 1.81 to 6.50, P < 0.01] compared with no erythromycin. In addition, erythromycin infusion before endoscopy resulted in a statistically significant decrease in the need for a second endoscopy (OR 0.47; 95% CI: 0.26 to 0.83, P = 0.01), units of blood transfused (WMD - 0.41; 95% CI: -0.82 to -0.01, P = 0.04), and the duration of hospital stay (WMD - 1.51; 95% CI: -2.45 to -0.56, P < 0.01). Erythromycin infusion before endoscopy in patients with UGIB significantly improves visualization of gastric mucosa while decreasing the need for a second endoscopy, units of blood transfused, and duration of hospital stay.
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard