- Indoor water and dampness and the health effects on children: a review.
Indoor water and dampness and the health effects on children: a review.
Current allergy and asthma reports (2013-11-20)
Kevin Kennedy, Carl Grimes
PMID24249387
ABSTRACT
Exposure to dampness and mold in indoor environments has received significant attention in recent research. This review focuses on studies looking at the impact of dampness, moisture and microbial agents on children's health. We then look more broadly at how research studies have tried to describe dampness and exposure to moisture. Papers published from 2010 onwards were reviewed and are briefly summarized. Most of the papers using dampness as a proxy for microbial exposure focused on respiratory tract infections, especially asthma. This review highlights new findings and also discusses the variety of approaches used to assess dampness in indoor environments.
Product Description
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
SKU
Pack Size
Availability
Price
Quantity
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C