MilliporeSigma
  • Modeling of hemophilia A using patient-specific induced pluripotent stem cells derived from urine cells.
Modeling of hemophilia A using patient-specific induced pluripotent stem cells derived from urine cells.
Life sciences (2014-05-20)
Bei Jia, Shen Chen, Zhiju Zhao, Pengfei Liu, Jinglei Cai, Dajiang Qin, Juan Du, Changwei Wu, Qianyu Chen, Xiujuan Cai, Hui Zhang, Yanhong Yu, Duanqing Pei, Mei Zhong, Guangjin Pan
ABSTRACT
Hemophilia A (HA) is a severe, congenital bleeding disorder caused by the deficiency of clotting factor VIII (FVIII). For years, traditional laboratory animals have been used to study HA and its therapies, although animal models may not entirely mirror the human pathophysiology. Human induced pluripotent stem cells (iPSCs) can undergo unlimited self-renewal and differentiate into all cell types. This study aims to generate hemophilia A (HA) patient-specific iPSCs that differentiate into disease-affected hepatocyte cells. These hepatocytes are potentially useful for in vitro disease modeling and provide an applicable cell source for autologous cell therapy after genetic correction. In this study, we mainly generated iPSCs from urine collected from HA patients with integration-free episomal vectors PEP4-EO2S-ET2K containing human genes OCT4, SOX2, SV40LT and KLF4, and differentiated these iPSCs into hepatocyte-like cells. We further identified the genetic phenotype of the FVIII genes and the FVIII activity in the patient-specific iPSC derived hepatic cells. HA patient-specific iPSCs (HA-iPSCs) exhibited typical pluripotent properties evident by immunostaining, in vitro assays and in vivo assays. Importantly, we showed that HA-iPSCs could differentiate into functional hepatocyte-like cells and the HA-iPSC-derived hepatocytes failed to produce FVIII, but otherwise functioned normally, recapitulating the phenotype of HA disease in vitro. HA-iPSCs, particular those generated from the urine using a non-viral approach, provide an efficient way for modeling HA in vitro. Furthermore, HA-iPSCs and their derivatives serve as an invaluable cell source that can be used for gene and cell therapy in regenerative medicine.
Product Description
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Human coagulation factor VIII concentrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Periodic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, crystallized, ≥99.0% (RT)
Sigma-Aldrich
Periodic acid, ACS reagent, 99%
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard