MilliporeSigma
  • Recovery of struvite via coagulation and flocculation using natural compounds.
Recovery of struvite via coagulation and flocculation using natural compounds.
Environmental technology (2014-08-26)
Maryam Latifian, Jing Liu, Bo Mattiasson
ABSTRACT
One of the major setbacks of struvite recovery processes is the difficulty in harvesting struvite crystals. This study evaluates the use of different coagulants to improve precipitation of struvite (MgNH4PO4.6H20) crystals. Chitosan and poly(diallyldimethyl ammonium chloride) (Poly-DADMAC) as a coagulant-flocculent and alginate and bentonite as a coagulant aid have been examined in jar tests. Also, a continuous three-phase process, i.e., struvite crystallization, coagulation/flocculation and precipitation process, was set up for real wastewater. Addition of chitosan as the coagulant and bentonite as the coagulant aid was significantly more efficient in forming struvite flocs in comparison to Poly-DADMAC alone or with coagulant aid, which did not show any positive effect. The calculated average settling velocity of struvite with chitosan-bentonite addition in synthetic and in real wastewater increased by approximately 5.3 and 2.8 folds, respectively, compared with that of no coagulant/flocculent addition. Phosphorus recovery of over 70% was achieved by the continuous process. Findings in this study clearly confirmed the possibility of using chitosan and bentonite as an efficient coagulant-flocculent to enhance the recovery of struvite crystals.
Sigma-Aldrich
Nanoclay, hydrophilic bentonite
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Bentonite, CP
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Bentonite