Sigma-Aldrich
Zinc chloride, AnhydroBeads™, amorphous, −10 mesh, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, AnhydroBeads™, amorphous, −10 mesh, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, JIS special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, SAJ first grade, ≥95.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride 0.1 M solution
Sign Into View Organizational & Contract Pricing Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sign Into View Organizational & Contract Pricing Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in ethanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide solution, 0.5 M in ethanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide solution, 0.02 M in ethanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Potassium hydroxide solution, 1 M
Sign Into View Organizational & Contract Pricing