MilliporeSigma
Synthetase polyspecificity as a tool to modulate protein function.
Bioorganic & medicinal chemistry letters (2011-11-02)
Douglas D Young, Steffen Jockush, Nicholas J Turro, Peter G Schultz
ABSTRACT
The site-specific incorporation of unnatural amino acids (UAAs) into proteins in bacteria is made possible by the evolution of aminoacyl-tRNA synthetases that selectively recognize and aminoacylate the amino acid of interest. Recently we have discovered that some of the previously evolved aaRSs display a degree of polyspecificity and are capable of recognizing multiple UAAs. Herein we report the polyspecificity of an aaRS evolved to encode a comarin containing amino acid. This polyspecificity was then exploited to introduce several UAAs into the fluorophore of GFP, altering its photophysical properties.
Sigma-Aldrich
H-4-(7-Hydroxy-4-coumarinyl)-Abu-OH, 98% (HPLC)