MilliporeSigma
  • Mutation of the conserved polyadenosine RNA binding protein, ZC3H14/dNab2, impairs neural function in Drosophila and humans.
Mutation of the conserved polyadenosine RNA binding protein, ZC3H14/dNab2, impairs neural function in Drosophila and humans.
Proceedings of the National Academy of Sciences of the United States of America (2011-07-08)
Changhui Pak, Masoud Garshasbi, Kimia Kahrizi, Christina Gross, Luciano H Apponi, John J Noto, Seth M Kelly, Sara W Leung, Andreas Tzschach, Farkhondeh Behjati, Seyedeh Sedigheh Abedini, Marzieh Mohseni, Lars R Jensen, Hao Hu, Brenda Huang, Sara N Stahley, Guanglu Liu, Kathryn R Williams, Sharon Burdick, Yue Feng, Subhabrata Sanyal, Gary J Bassell, Hans-Hilger Ropers, Hossein Najmabadi, Anita H Corbett, Kenneth H Moberg, Andreas W Kuss
ABSTRACT
Here we report a human intellectual disability disease locus on chromosome 14q31.3 corresponding to mutation of the ZC3H14 gene that encodes a conserved polyadenosine RNA binding protein. We identify ZC3H14 mRNA transcripts in the human central nervous system, and we find that rodent ZC3H14 protein is expressed in hippocampal neurons and colocalizes with poly(A) RNA in neuronal cell bodies. A Drosophila melanogaster model of this disease created by mutation of the gene encoding the ZC3H14 ortholog dNab2, which also binds polyadenosine RNA, reveals that dNab2 is essential for development and required in neurons for normal locomotion and flight. Biochemical and genetic data indicate that dNab2 restricts bulk poly(A) tail length in vivo, suggesting that this function may underlie its role in development and disease. These studies reveal a conserved requirement for ZC3H14/dNab2 in the metazoan nervous system and identify a poly(A) RNA binding protein associated with a human brain disorder.
Sigma-Aldrich
Anti-ZC3H14 antibody produced in rabbit, affinity isolated antibody
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$541.00