MilliporeSigma
  • The corepressor CTBP2 is a coactivator of retinoic acid receptor/retinoid X receptor in retinoic acid signaling.
The corepressor CTBP2 is a coactivator of retinoic acid receptor/retinoid X receptor in retinoic acid signaling.
Molecular and cellular biology (2013-06-19)
Prashanth Kumar Bajpe, Guus J J E Heynen, Lorenza Mittempergher, Wipawadee Grernrum, Iris A de Rink, Wouter Nijkamp, Roderick L Beijersbergen, Rene Bernards, Sidong Huang
ABSTRACT
Retinoids play key roles in development, differentiation, and homeostasis through regulation of specific target genes by the retinoic acid receptor/retinoid X receptor (RAR/RXR) nuclear receptor complex. Corepressors and coactivators contribute to its transcriptional control by creating the appropriate chromatin environment, but the precise composition of these nuclear receptor complexes remains to be elucidated. Using an RNA interference-based genetic screen in mouse F9 cells, we identified the transcriptional corepressor CTBP2 (C-terminal binding protein 2) as a coactivator critically required for retinoic acid (RA)-induced transcription. CTBP2 suppression by RNA interference confers resistance to RA-induced differentiation in diverse murine and human cells. Mechanistically, we find that CTBP2 associates with RAR/RXR at RA target gene promoters and is essential for their transactivation in response to RA. We show that CTBP2 is indispensable to create a chromatin environment conducive for RAR/RXR-mediated transcription by recruiting the histone acetyltransferase p300. Our data reveal an unexpected function of the corepressor CTBP2 as a coactivator for RAR/RXR in RA signaling.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Leukocyte Alkaline Phosphatase Kit, based on naphthol AS-BI and fast red violet LB
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit