MilliporeSigma
  • The effects of acrylamide polyelectrolytes on aquatic organisms: relating toxicity to chain architecture.
The effects of acrylamide polyelectrolytes on aquatic organisms: relating toxicity to chain architecture.
Chemosphere (2014-07-23)
R Costa, J L Pereira, J Gomes, F Gonçalves, D Hunkeler, M G Rasteiro
ABSTRACT
Understanding the inherent toxicity of water-soluble synthetic polyelectrolytes is critical for adequate risk management as well as enhancing product design when biological activity is a key performance index (e.g. for application in biofouling bivalves' control). The toxicity of two cationic acrylamide copolymers with different chain branching degree was evaluated. Standard ecotoxicity tests were conducted with microalgae and daphnids. The susceptibility of Corbicula fluminea, as a biofouling bivalve, was also evaluated. The effect of polyelectrolyte on the test media viscosity and the polymer chain size distributions under the experimental conditions were also examined. The susceptibility of the microalgae to both polymers was similar. As the complexity and size of the test organisms increased, differences in toxicity due to different chain architecture were noticeable. The more branched polymer was significantly less toxic to both daphnids and the bivalves, which could be linked to the distinctive features of its bimodal size chain distribution. This architecture resulted in both more compact globular molecules and the formation of aggregates, which reduce the polymer interaction with the biological surfaces. The results of this study promote the incorporation of environmental considerations in polyelectrolyte development and contribute to the design of improved solutions for controlling biofouling bivalves.
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sign Into View Organizational & Contract Pricing
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
Supelco
Acrylamide, analytical standard
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Distilled water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
Supelco
Water, for TOC analysis
Sign Into View Organizational & Contract Pricing
Supelco
Water, suitable for ion chromatography
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
Supelco
Water, ACS reagent, for ultratrace analysis
Sign Into View Organizational & Contract Pricing
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, ACS reagent
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, Deionized
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, HPLC Plus
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Acrylamide, ≥99.9%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Acrylamide, ≥98.0%
Sign Into View Organizational & Contract Pricing