MilliporeSigma
  • Positive symptoms and water diffusivity of the prefrontal and temporal cortices in schizophrenia patients: a pilot study.
Positive symptoms and water diffusivity of the prefrontal and temporal cortices in schizophrenia patients: a pilot study.
Psychiatry research (2014-08-12)
Jin Young Park, Hae-Jeong Park, Dae-Jin Kim, Jae-Jin Kim
ABSTRACT
The development of diffusion tensor imaging (DTI) has provided information about microstructural changes in the brain. Most DTI studies have focused on white matter (WM). Few DTI studies have examined the gray matter (GM) in schizophrenia and, to date, there has been no attempt to identify the relationship between water diffusivity and symptom severity in schizophrenia. The present study aimed to examine microstructural deficits in the dorsal prefrontal cortex (DPFC) and temporal cortex in schizophrenia patients using fractional anisotropy (FA) and water diffusivity. This study also explored the relationship between DTI measurements and psychotic symptoms. Magnetic resonance imaging (MRI) and DTI were used to study 19 schizophrenia patients and 19 healthy controls. Fractional anisotropy, axial diffusivity, radial diffusivity, and regional volumes were measured in the prefrontal cortex and temporal cortex. On DTI measurements, patients showed increased axial and radial diffusivities in the prefrontal cortex and temporal cortex, but they did not demonstrate any difference in fractional anisotropy and regional volumes. Additionally, axial and radial diffusivities were significantly correlated with positive symptom scores in all regions of interest. These results indicate that water diffusivity measurements, including axial and radial diffusivities, can be used to identify microstructural changes in the gray matter in schizophrenia that may be related to symptom severity.
Product Description
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Distilled water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
Water, for TOC analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
Water, suitable for ion chromatography
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Supelco
Water, ACS reagent, for ultratrace analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, Deionized
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00
Sigma-Aldrich
Water, HPLC Plus
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
In Stock
$429.00