MilliporeSigma
  • Interleukin 22 protects colorectal cancer cells from chemotherapy by activating the STAT3 pathway and inducing autocrine expression of interleukin 8.
Interleukin 22 protects colorectal cancer cells from chemotherapy by activating the STAT3 pathway and inducing autocrine expression of interleukin 8.
Clinical immunology (Orlando, Fla.) (2014-07-27)
Tingyu Wu, Zhongchuan Wang, Yun Liu, Zubing Mei, Guanghui Wang, Zhonglin Liang, Ang Cui, Xuguang Hu, Long Cui, Yili Yang, Chen-Ying Liu
ABSTRACT
Resistance to chemotherapy is the major cause of colorectal cancer (CRC) treatment failure. The cytokine IL-22, which is produced by T cells and NK cells, is associated with tumorigenesis and tumor progression in cancers. However, the role of IL-22 in chemoresistance has not been investigated. We found that IL-22 levels in tumor tissues and peripheral blood were associated with chemoresistance and indicate poor prognosis for patients who received FOLFOX chemotherapy. In CRC cells, IL-22 was able to attenuate the cytotoxic and apoptosis-inducing effects of 5-FU and OXA by activating the STAT3 pathway and subsequently increasing the expression of anti-apoptotic genes. In addition, IL-22 conferred resistance to 5-FU and OXA by inducing IL-8 autocrine expression through STAT3 activation. Our findings identify IL-22 as a novel chemoresistance cytokine and may be a useful prognostic biomarker for CRC patients receiving FOLFOX chemotherapy.
Sigma-Aldrich
IL-22 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Human IL-22 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
IL-22 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Supelco
5-Fluorouracil, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Folinic acid calcium salt hydrate, BioXtra, ≥99.0% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Supelco
Folinic acid calcium salt hydrate, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Sigma-Aldrich
Oxaliplatin, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00
Calcium folinate, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Estimated to ship on March 24, 2025
$150.00