MilliporeSigma
  • Insight into the stability of poly(diallydimethylammoniumchloride) and polybrene poly cationic coatings in capillary electrophoresis.
Insight into the stability of poly(diallydimethylammoniumchloride) and polybrene poly cationic coatings in capillary electrophoresis.
Journal of chromatography. A (2014-09-28)
Lei Pei, Charles A Lucy
ABSTRACT
Polycationic polymers are widely used in capillary electrophoresis (CE) as surface coatings to prevent protein adsorption and control electroosmotic flow (EOF). Such semi-permanent coatings are formed by flushing the capillary with a quaternary amine-based polymer such as poly(diallydimethylammonium chloride) (PDADMAC) or polybrene. Compared to covalent capillary coatings, the claimed advantages of adsorptive polycation coatings are their simple preparation and that they are not limited to the pH 2-8 range as are covalent coatings. However, while the latter is commonly claimed, few studies have demonstrated the stability of polycationic coatings at extreme pH. Herein PDADMAC and polybrene are studied as model cationic coatings. PDADMAC with higher molecular weight (M.W.) demonstrated higher EOF stability at pH 9.5, with PDADMAC of M.W. less than 200,000 being unstable at pH 9.5. X-ray photoelectron spectroscopy (XPS) shows that the quaternary amines of PDADMAC and polybrene were slowly converted to tertiary amines in alkaline solution and more rapidly when adsorbed on a silica surface. The degraded polycation deprotonated at pH >7, resulting in loss of polymer from the surface and diminishing EOF. Successive multiple ionic layer (SMIL) coatings show greater alkaline stability by distancing the polycation from the surface. Separations of inorganic anions at pH 9.5 illustrate the degradation behavior and enhanced stability of higher M.W. polycationic coatings.
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O