MilliporeSigma
  • Enantioselective cyclization of enamide-ynes and application to the synthesis of the kopsifoline core.
Enantioselective cyclization of enamide-ynes and application to the synthesis of the kopsifoline core.
Tetrahedron (2013-06-19)
Britton K Corkey, Stephen T Heller, Yi-Ming Wang, F Dean Toste
ABSTRACT
We report the palladium-catalyzed enantioselective cyclization of 1,6-enamidynes to form spirocyclic ring systems. We applied this methodology to the concise synthesis of the skeletal core of the kopsifoline alkaloids.
Product Description
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 20 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 50 m × 0.25 mm, df 0.12 μm