MilliporeSigma
  • Identification of a basal-like subtype of breast ductal carcinoma in situ.
Identification of a basal-like subtype of breast ductal carcinoma in situ.
Human pathology (2007-01-20)
Chad A Livasy, Charles M Perou, Gamze Karaca, David W Cowan, Diane Maia, Susan Jackson, Chiu-Kit Tse, Sarah Nyante, Robert C Millikan
ABSTRACT
Microarray profiling of invasive breast carcinomas has identified subtypes including luminal A, luminal B, HER2-overexpressing, and basal-like. The poor-prognosis, basal-like tumors have been immunohistochemically characterized as estrogen receptor (ER)-negative, HER2/neu-negative, and cytokeratin 5/6-positive and/or epidermal growth factor receptor (EGFR)-positive. The aim of this study was to determine the prevalence of basal-like ductal carcinoma in situ in a population-based series of cases using immunohistochemical surrogates. A total of 245 pure ductal carcinoma in situ cases from a population-based, case-control study were evaluated for histologic characteristics and immunostained for ER, HER2/neu, EGFR, cytokeratin 5/6, p53, and Ki-67. The subtypes were defined as: luminal A (ER+, HER2-), luminal B (ER+, HER2+), HER2 positive (ER-, HER2+), and basal-like (ER-, HER2-, EGFR+, and/or cytokeratin 5/6+). The prevalence of breast cancer subtypes was basal-like (n = 19 [8%]); luminal A, n = 149 (61%); luminal B, n = 23 (9%); and HER2+/ER-, n = 38 (16%). Sixteen tumors (6%) were unclassified (negative for all 4 defining markers). The basal-like subtype was associated with unfavorable prognostic variables including high-grade nuclei (P < .0001), p53 overexpression (P < .0001), and elevated Ki-67 index (P < .0001). These studies demonstrate the presence of a basal-like in situ carcinoma, a potential precursor lesion to invasive basal-like carcinoma.
Sigma-Aldrich
Cytokeratin 5 (EP1601Y) + Cytokeratin 14 (LL002), Mouse and Rabbit Monoclonal Antibody
Sigma-Aldrich
Cytokeratin 5 (EP1601Y) Rabbit Monoclonal Primary Antibody