MilliporeSigma
  • Electroconvulsive therapy augmentation in clozapine-resistant schizophrenia: a prospective, randomized study.
Electroconvulsive therapy augmentation in clozapine-resistant schizophrenia: a prospective, randomized study.
The American journal of psychiatry (2014-08-27)
Georgios Petrides, Chitra Malur, Raphael J Braga, Samuel H Bailine, Nina R Schooler, Anil K Malhotra, John M Kane, Sohag Sanghani, Terry E Goldberg, Majnu John, Alan Mendelowitz
ABSTRACT
Up to 70% of patients with treatment-resistant schizophrenia do not respond to clozapine. Pharmacological augmentation to clozapine has been studied with unimpressive results. The authors examined the use of ECT as an augmentation to clozapine for treatment-refractory schizophrenia. In a randomized single-blind 8-week study, patients with clozapine-resistant schizophrenia were assigned to treatment as usual (clozapine group) or a course of bilateral ECT plus clozapine (ECT plus clozapine group). Nonresponders from the clozapine group received an 8-week open trial of ECT (crossover phase). ECT was performed three times per week for the first 4 weeks and twice weekly for the last 4 weeks. Clozapine dosages remained constant. Response was defined as ≥40% reduction in symptoms based on the psychotic symptom subscale of the Brief Psychiatric Rating Scale, a Clinical Global Impressions (CGI)-severity rating <3, and a CGI-improvement rating ≤2. The intent-to-treat sample included 39 participants (ECT plus clozapine group, N=20; clozapine group, N=19). All 19 patients from the clozapine group received ECT in the crossover phase. Fifty percent of the ECT plus clozapine patients met the response criterion. None of the patients in the clozapine group met the criterion. In the crossover phase, response was 47%. There were no discernible differences between groups on global cognition. Two patients required the postponement of an ECT session because of mild confusion. The augmentation of clozapine with ECT is a safe and effective treatment option. Further research is required to determine the persistence of the improvement and the potential need for maintenance treatments.
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Clozapine
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
USP
Clozapine, United States Pharmacopeia (USP) Reference Standard
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Clozapine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Clozapine for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
USP
Clozapine Resolution Mixture, United States Pharmacopeia (USP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Caffeine, European Pharmacopoeia (EP) Reference Standard