MilliporeSigma
  • Phenylephrine enhances glutamate release in the medial prefrontal cortex through interaction with N-type Ca2+ channels and release machinery.
Phenylephrine enhances glutamate release in the medial prefrontal cortex through interaction with N-type Ca2+ channels and release machinery.
Journal of neurochemistry (2014-09-10)
Fei Luo, Si-Hai Li, Hua Tang, Wei-Ke Deng, Yu Zhang, Ying Liu
ABSTRACT
α1 -adrenoceptors (α1 -ARs) stimulation has been found to enhance excitatory processes in many brain regions. A recent study in our laboratory showed that α1 -ARs stimulation enhances glutamatergic transmission via both pre- and post-synaptic mechanisms in layer V/VI pyramidal cells of the rat medial prefrontal cortex (mPFC). However, a number of pre-synaptic mechanisms may contribute to α1 -ARs-induced enhancement of glutamate release. In this study, we blocked the possible post-synaptic action mediated by α1 -ARs to investigate how α1 -ARs activation regulates pre-synaptic glutamate release in layer V/VI pyramidal neurons of mPFC. We found that the α1 -ARs agonist phenylephrine (Phe) induced a significant enhancement of glutamatergic transmission. The Phe-induced potentiation was mediated by enhancing pre-synaptic glutamate release probability and increasing the number of release vesicles via a protein kinase C-dependent pathway. The mechanisms of Phe-induced potentiation included interaction with both glutamate release machinery and N-type Ca(2+) channels, probably via a pre-synaptic Gq /phospholipase C/protein kinase C pathway. Our results may provide a cellular and molecular mechanism that helps explain α1 -ARs-mediated influence on PFC cognitive functions. Alpha1 -adrenoceptor (α1 -ARs) stimulation has been reported to enhance glutamatergic transmission in layer V/VI pyramidal neurons of the rat medial prefrontal cortex (mPFC). We found that α1 -ARs agonist phenylephrine (Phe) increases pre-synaptic glutamate release probability and the number of released vesicles via interaction with both glutamate release machinery and N-type Ca(2+) channels. Our results may provide a cellular and molecular mechanism that helps explain α1 -ARs-mediated influence on PFC cognitive functions. Gq, Gq protein; PLC, phospholipase C; PKC, protein kinase C; AMPA, α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid; NMDA, N-methyl-d-aspartate; Glu, glutamate; Phe, phenylephrine.
Product Description
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid hydrochloride
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, JIS special grade, ≥99.0%
Supelco
R-(-)-Phenylephrine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Phenylephrine, European Pharmacopoeia (EP) Reference Standard