MilliporeSigma
  • Outcome of 3-day bleomycin, etoposide and cisplatin chemotherapeutic regimen for patients with malignant ovarian germ cell tumours: a Taiwanese Gynecologic Oncology Group study.
Outcome of 3-day bleomycin, etoposide and cisplatin chemotherapeutic regimen for patients with malignant ovarian germ cell tumours: a Taiwanese Gynecologic Oncology Group study.
European journal of cancer (Oxford, England : 1990) (2014-12-03)
Chi-An Chen, Hao Lin, Chia-Sui Weng, Kuo-Chang Wen, Chien-Hsing Lu, Hung-Hsueh Chou, Yu-Fang Huang, Chieh-Yi Kang, Chih-Ming Ho, Mu-Hsien Yu, Cheng-Yang Chou
ABSTRACT
The combination of bleomycin, etoposide and cisplatin (BEP) is currently the most widely used treatment for malignant ovarian germ cell tumours (MOGCTs). The aim of this study was to evaluate the efficacy and adverse effects of the 3-day BEP regimen in Taiwan. The prognostic factors of the MOGCT patients were also analysed. Two hundred and thirty-nine cases of MOGCTs were identified from the Taiwanese Gynecologic Oncology Group database, and 204 of those who received postoperative BEP chemotherapy were then analysed. The estimated rate of no evidence of disease was 94.0% for 204 patients with adjuvant BEP regimen. Seven grade 3/4 haematological adverse effects including four subjects with neutropenia, one with pancytopenia and two with neutropenic fever were recorded in the 853 total courses of chemotherapeutic cycles. The rates of haematological and non-haematological adverse effects were 0.82% and 2.3%, respectively. No treatment-related mortality was noted. In the analysis of prognostic factors, only tumour stage had a significant impact on disease recurrence (95% confidence interval (CI), 4.2–94.4, p < 0.001) and disease-related mortality (95% CI, 2.2–163.9, p = 0.007). The current 3-day adjuvant BEP regimen was effective and safe for patients with MOGCTs.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard