MilliporeSigma
  • On the time course, generality, and regulation of plasma progesterone release in male rats by stress exposure.
On the time course, generality, and regulation of plasma progesterone release in male rats by stress exposure.
Endocrinology (2014-06-14)
Cara M Hueston, Terrence Deak
ABSTRACT
Although progesterone is most commonly regarded in terms of its role in the female estrous cycle, reproductive behavior, and pregnancy, progesterone is also a precursor to corticosterone (CORT) and is released from the adrenal glands of both sexes in response to stress. However, the relationship between plasma CORT and progesterone during times of stress has not been well established. To better characterize dynamic changes in progesterone release as a result of stressor exposure, plasma progesterone levels were measured using enzyme immunoassay under multiple conditions, including after stress exposure (footshock, restraint, and forced swim), manipulations of the hypothalamic-pituitary-adrenal axis (injection of metyrapone or dexamethasone, or adrenalectomy), and in response to CRH and ACTH injections. When plasma levels of ACTH, CORT, and progesterone were analyzed across studies, CORT and progesterone were found to be highly correlated and rarely dissociated. Additionally, it appears that in male rats, the adrenal glands are the principle source of circulating progesterone in response to stress. Interestingly, a detailed time course indicated lack of a circadian rhythm of progesterone secretion, despite a robust rhythm of circulating CORT. The current studies provide critical new information on the coincident release of progesterone and CORT in response to stress and their somewhat paradoxical dissociation across the circadian rhythm. These findings provide an important foundation for future studies that will examine the role of stress-induced progesterone in behavioral, neuroimmune, and neuroendocrine responses to stress.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis