MilliporeSigma
  • BclAF1 restriction factor is neutralized by proteasomal degradation and microRNA repression during human cytomegalovirus infection.
BclAF1 restriction factor is neutralized by proteasomal degradation and microRNA repression during human cytomegalovirus infection.
Proceedings of the National Academy of Sciences of the United States of America (2012-05-31)
Song Hee Lee, Robert F Kalejta, Julie Kerry, Oliver John Semmes, Christine M O'Connor, Zia Khan, Benjamin A Garcia, Thomas Shenk, Eain Murphy
ABSTRACT
Cell proteins can restrict the replication of viruses. Here, we identify the cellular BclAF1 protein as a human cytomegalovirus restriction factor and describe two independent mechanisms the virus uses to decrease its steady-state levels. Immediately following infection, the viral pp71 and UL35 proteins, which are delivered to cells within virions, direct the proteasomal degradation of BclAF1. Although BclAF1 reaccumulates through the middle stages of infection, it is subsequently down-regulated at late times by miR-UL112-1, a virus-encoded microRNA. In the absence of BclAF1 neutralization, viral gene expression and replication are inhibited. These data identify two temporally and mechanistically distinct functions used by human cytomegalovirus to down-regulate a cellular antiviral protein.
Sigma-Aldrich
Anti-Daxx antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)