MilliporeSigma
  • Establishing the stability and reversibility of protein pyrophosphorylation with synthetic peptides.
Establishing the stability and reversibility of protein pyrophosphorylation with synthetic peptides.
Chembiochem : a European journal of chemical biology (2015-02-03)
Lisa M Yates, Dorothea Fiedler
ABSTRACT
Protein pyrophosphorylation is emerging as a new post-translational modification, yet its role in cellular signaling remains poorly characterized. Important factors in determining the biological relevance of pyrophosphorylation include understanding the chemical and biochemical stability of the pyrophosphoryl group and elucidating the reversibility of modification in a cellular context. Towards this end, we prepared a series of synthetic pyrophosphopeptides, which were utilized to demonstrate that the modification is quite inert over a wide pH range but can be removed biochemically by alkaline phosphatases. Importantly, we observed enzyme-dependent removal of the pyrophosphate in mammalian and yeast cell lysates using the synthetic pyrophosphopeptides. The findings provide evidence for the reversibility of pyrophosphorylation and thereby highlight the potential impact of this modification on cellular signal transduction pathways.
Product Description
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
SKU
Pack Size
Availability
Price
Quantity
Supelco
Water, for TOC analysis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Water, suitable for ion chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for molecular biology, sterile filtered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
E-Toxate Water, endotoxin, free
SKU
Pack Size
Availability
Price
Quantity
Supelco
Water, ACS reagent, for ultratrace analysis
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
SKU
Pack Size
Availability
Price
Quantity
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%