MilliporeSigma
  • Cationic core-shell nanoparticles for intravesical chemotherapy in tumor-induced rat model: safety and efficacy.
Cationic core-shell nanoparticles for intravesical chemotherapy in tumor-induced rat model: safety and efficacy.
International journal of pharmaceutics (2014-05-20)
Nazlı Erdogar, Alper B İskit, Hakan Eroglu, Mustafa F Sargon, N Aydın Mungan, Erem Bilensoy
ABSTRACT
Mitomycin C (MMC) has shown potent efficacy against a wide spectrum of cancers and is clinical first choice in superficial bladder tumors. However, intravesical chemotherapy with MMC has been ineffective due to periodical discharge of the bladder and instability of this drug in acidic pH, both resulting in high rate of tumor recurrence and insufficiency to prevent progression. Nanocarriers may be a promising alternative for prolonged, effective and safe intravesical drug delivery due to their favorable size, surface properties and optimum interaction with mucosal layer of the bladder wall. Hence, the aim of this study was to evaluate and optimize cationic core-shell nanoparticles formulations (based on chitosan (CS) and poly-ϵ-caprolactone (PCL)) in terms of antitumor efficacy after intravesical administration in bladder tumor induced rat model. Antitumor efficacy was determined through the parameters of survival rate and nanoparticle penetration into the bladder tissue. Safety of the formulations were evaluated by histopathological evaluation of bladder tissue as well as observation of animals treated with MMC bound to nanoparticles. Results indicated that chitosan coated poly-ϵ-caprolactone (CS-PCL) nanoparticles presented the longest survival rate among all treatment groups as evaluated by Kaplan-Meier plotting. Histopathological evaluation revealed that cationic nanoparticles were localized and accumulated in the bladder tissue. As intravesical chemotherapy is a local therapy, no MMC was quantified in blood after intravesical instillation indicating no systemic uptake for the drug which could have subsequently led to side effects. In conclusion, core-shell type cationic nanoparticles may be effective tools for the intravesical chemotherapy of recurrent bladder tumors.
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, Selectophore, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)