MilliporeSigma
  • The promotion of a constructive macrophage phenotype by solubilized extracellular matrix.
The promotion of a constructive macrophage phenotype by solubilized extracellular matrix.
Biomaterials (2014-07-22)
Brian M Sicari, Jenna L Dziki, Bernard F Siu, Christopher J Medberry, Christopher L Dearth, Stephen F Badylak
ABSTRACT
The regenerative healing response of injured skeletal muscle is dependent upon a heterogeneous population of responding macrophages, which show a phenotypic transition from the pro-inflammatory M1 to the alternatively activated and constructive M2 phenotype. Biologic scaffolds derived from mammalian extracellular matrix (ECM) have been used for the repair and reconstruction of a variety of tissues, including skeletal muscle, and have been associated with an M2 phenotype and a constructive and functional tissue response. The mechanism(s) behind in-vivo macrophage phenotype transition in skeletal muscle and the enhanced M2:M1 ratio associated with ECM bioscaffold use in-vivo are only partially understood. The present study shows that degradation products from ECM bioscaffolds promote alternatively activated and constructive M2 macrophage polarization in-vitro, which in turn facilitates migration and myogenesis of skeletal muscle progenitor cells.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)