MilliporeSigma
  • The dismantling of Calliobothrium (Cestoda: Tetraphyllidea) with erection of Symcallio n. gen. and description of two new species.
The dismantling of Calliobothrium (Cestoda: Tetraphyllidea) with erection of Symcallio n. gen. and description of two new species.
The Journal of parasitology (2014-12-17)
James P Bernot, Janine N Caira, Maria Pickering
ABSTRACT
This paper aims to resolve the dual composition of the triakid shark-hosted tetraphyllidean genus Calliobothrium--an issue that has been recognized for over a decade. As it stands, this genus includes a number of large species with laciniate proglottids, most of which bear 3 suckers at the anterior margin of each bothridium, but it also includes a number of species that lack proglottid laciniations and bear only a single sucker per bothridium, most of which are relatively small. Discovery of 2 new species, 1 of each form, parasitizing the whitespot smoothhound shark, Mustelus palumbes, off South Africa, prompted the first molecular analysis of the genus. Bayesian inference and maximum likelihood analyses of 28S rDNA (D1-D3) sequence data generated for specimens of both new species as well as of 2 known species of the laciniate form (Calliobothrium australis and Calliobothrium cf. verticillatum) and 3 known species of the non-laciniate form (Calliobothrium violae, Calliobothrium riseri, and Calliobothrium barbarae) confirmed the reciprocal monophyly of the 2 clades, supporting establishment of a new genus. Because the type of Calliobothrium, C. verticillatum, is of the laciniate form, Symcallio n. gen., with Symcallio peteri n. gen., n. sp., from M. palumbes described as its type, is established to house members of the non-laciniate clade. The 11 described species consistent with this form are transferred to the new genus. A new species of the laciniate clade, Calliobothrium euzeti n. sp., is described from M. palumbes, and a revised diagnosis of Calliobothrium is presented.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
o-Xylene, analytical standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
o-Xylene, suitable for HPLC, 98%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Ethanol, ≥99.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.