MilliporeSigma
  • Local inhibition of 5-lipoxygenase enhances bone formation in a rat model.
Local inhibition of 5-lipoxygenase enhances bone formation in a rat model.
Bone & joint research (2013-04-24)
J A Cottrell, V Keshav, A Mitchell, J P O'Connor
ABSTRACT
Recent studies have shown that modulating inflammation-related lipid signalling after a bone fracture can accelerate healing in animal models. Specifically, decreasing 5-lipoxygenase (5-LO) activity during fracture healing increases cyclooxygenase-2 (COX-2) expression in the fracture callus, accelerates chondrogenesis and decreases healing time. In this study, we test the hypothesis that 5-LO inhibition will increase direct osteogenesis. Bilateral, unicortical femoral defects were used in rats to measure the effects of local 5-LO inhibition on direct osteogenesis. The defect sites were filled with a polycaprolactone (PCL) scaffold containing 5-LO inhibitor (A-79175) at three dose levels, scaffold with drug carrier, or scaffold only. Drug release was assessed in vitro. Osteogenesis was assessed by micro-CT and histology at two endpoints of ten and 30 days. Using micro-CT, we found that A-79175, a 5-LO inhibitor, increased bone formation in an apparent dose-related manner. These results indicate that 5-LO inhibition could be used therapeutically to enhance treatments that require the direct formation of bone.
Product Description
Supelco
Water, for TOC analysis
Pricing and availability is not currently available.
Supelco
Water, suitable for ion chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Water, BioPerformance Certified
Pricing and availability is not currently available.
Supelco
Water, ACS reagent, for ultratrace analysis
Pricing and availability is not currently available.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Pricing and availability is not currently available.
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pricing and availability is not currently available.
Sigma-Aldrich
Water, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Pricing and availability is not currently available.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Water, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Water, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Water, HPLC Plus
Pricing and availability is not currently available.
Sigma-Aldrich
Water, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Deionized
Pricing and availability is not currently available.
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pricing and availability is not currently available.
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pricing and availability is not currently available.