MilliporeSigma
  • IL-15 induces alloreactive CD28(-) memory CD8 T cell proliferation and CTLA4-Ig resistant memory CD8 T cell activation.
IL-15 induces alloreactive CD28(-) memory CD8 T cell proliferation and CTLA4-Ig resistant memory CD8 T cell activation.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2014-05-21)
O Traitanon, A Gorbachev, J J Bechtel, K S Keslar, W M Baldwin, E D Poggio, R L Fairchild
ABSTRACT
The presence of CD28(-) memory CD8 T cells in the peripheral blood of renal transplant patients is a risk factor for graft rejection and resistance to CTLA-4Ig induction therapy. In vitro analyses have indicated poor alloantigen-induced CD28(-) memory CD8 T cell proliferation, raising questions about mechanisms mediating their clonal expansion in kidney grafts to mediate injury. Candidate proliferative cytokines were tested for synergy with alloantigen in stimulating CD28(-) memory CD8 T cell proliferation. Addition of IL-15, but not IL-2 or IL-7, to co-cultures of CD28(-) or CD28(+) memory CD8 T cells and allogeneic B cells rescued proliferation of the CD28(-) and enhanced CD28(+) memory T cell proliferation. Proliferating CD28(-) memory CD8 T cells produced high amounts of interferon gamma and tumor necrosis factor alpha and expressed higher levels of the cytolytic marker CD107a than CD28(+) memory CD8 T cells. CTLA-4Ig inhibited alloantigen-induced proliferation of CD28(+) memory CD8 T cell proliferation but had no effect on alloantigen plus IL-15-induced proliferation of either CD28(-) or CD28(+) memory CD8 T cells. These results indicate the ability of IL-15, a cytokine produced by renal epithelial during inflammation, to provoke CD28(-) memory CD8 T cell proliferation and to confer memory CD8 T cell resistance to CTLA-4Ig-mediated costimulation blockade.
Sigma-Aldrich
Interleukin-7 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-15 human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-7 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-15 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture