MilliporeSigma
  • Association between some endocrine-disrupting chemicals and childhood obesity in biological samples of young girls: a cross-sectional study.
Association between some endocrine-disrupting chemicals and childhood obesity in biological samples of young girls: a cross-sectional study.
Environmental toxicology and pharmacology (2014-06-09)
Jaeyeon Choi, Jinhee Eom, Jiye Kim, Sanghouck Lee, Yunje Kim
ABSTRACT
Childhood obesity, a major public health concern, has increased worldwide. Endocrine-disrupting chemicals (EDCs) have recently received attention as a cause of obesity. A cross-sectional study using logistic regression was conducted to investigate the association between some endocrine disrupting chemicals and obesity in young girls. Endogenous steroids expected to be associated with EDCs were also investigated. The target compounds included 7 phthalates (MEP, DBP, MBP, DEHP, MEHP, PA and MBzP), 2 alkylphenols (4-NP and t-OP), bisphenol A and 9 endogenous steroids (DHT, epi-T, T, DHEA, A, P, E1, E2 and E3). PA in urine and MEP, DBP and PA in serum showed statistically significant differences between the control and obese groups, those compounds were considered to be associated with obesity. In addition, DHEA in serum showed a statistically significant difference between obese and control groups. We concluded that these substances can affect the development of obesity.
Product Description
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Supelco
Benzyl butyl phthalate, analytical standard
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Terephthalic acid, 98%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzyl butyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Supelco
Chlorotrimethylsilane, derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Terephthalic acid, analytical standard
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol, British Pharmacopoeia (BP) Assay Standard
Supelco
Metolachlor OA, PESTANAL®, analytical standard
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide, derivatization grade (GC derivatization), LiChropur, ≥99.0%