MilliporeSigma
  • Effector CD4 T-cell transition to memory requires late cognate interactions that induce autocrine IL-2.
Effector CD4 T-cell transition to memory requires late cognate interactions that induce autocrine IL-2.
Nature communications (2014-11-06)
K Kai McKinstry, Tara M Strutt, Bianca Bautista, Wenliang Zhang, Yi Kuang, Andrea M Cooper, Susan L Swain
ABSTRACT
It is unclear how CD4 T-cell memory formation is regulated following pathogen challenge, and when critical mechanisms act to determine effector T-cell fate. Here, we report that following influenza infection most effectors require signals from major histocompatibility complex class II molecules and CD70 during a late window well after initial priming to become memory. During this timeframe, effector cells must produce IL-2 or be exposed to high levels of paracrine or exogenously added IL-2 to survive an otherwise rapid default contraction phase. Late IL-2 promotes survival through acute downregulation of apoptotic pathways in effector T cells and by permanently upregulating their IL-7 receptor expression, enabling IL-7 to sustain them as memory T cells. This new paradigm defines a late checkpoint during the effector phase at which cognate interactions direct CD4 T-cell memory generation.
Product Description
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar