Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Imidazole, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri™
Sign Into View Organizational & Contract Pricing Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri™, free-flowing
Sign Into View Organizational & Contract Pricing Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99%
Sign Into View Organizational & Contract Pricing Supelco
Codeine monohydrate, analytical standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sign Into View Organizational & Contract Pricing Supelco
Ethyl acetate, analytical standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sign Into View Organizational & Contract Pricing