MilliporeSigma
  • Optimization of methyl jasmonate application to broccoli florets to enhance health-promoting phytochemical content.
Optimization of methyl jasmonate application to broccoli florets to enhance health-promoting phytochemical content.
Journal of the science of food and agriculture (2013-12-18)
Kang Mo Ku, Elizabeth H Jeffery, John A Juvik
ABSTRACT
Spray treatment of methyl jasmonate (MeJA) has been shown to increase glucosinolate (GS) concentrations and health-promoting activity in Brassica vegetables. Since there is no reported standardized protocol, several MeJA treatment studies have been conducted to maximize human health bioactivity using the F1 broccoli cultivar 'Green Magic'. Foliar MeJA application 4 days prior to harvest of broccoli at commercial maturity resulted in enhanced total GS concentrations. Although a single application of 250 µmol L(-1) MeJA maximized GS concentrations in broccoli florets, two days of consecutive treatments (4 and 3 days prior to harvest) of 250 µmol L(-1) MeJA further enhanced neoglucobrassicin concentrations and floret extract quinone reductase (QR)-inducing activity. With increasing concentrations of MeJA in spray applications to broccoli florets, concentrations of the glucosinolates glucoraphanin, gluconasturtiin and neoglucobrassicin and the isothiocyanate sulforaphane as well as anticancer and anti-inflammatory bioactivities as measured by QR induction and inhibition of nitric oxide (NO) production respectively were significantly increased. Concentrations of these phytochemicals showed strong positive correlations with QR-inducing and NO-inhibitory activities. These application protocols were found to maximize GS and GS hydrolysis product concentrations and putatively enhance the health-promoting properties of broccoli heads for consumers.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Kaempferol, analytical standard
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules