MilliporeSigma
  • Differential expression of CYP1A1 and CYP1A2 genes in H4IIE rat hepatoma cells exposed to TCDD and PAHs.
Differential expression of CYP1A1 and CYP1A2 genes in H4IIE rat hepatoma cells exposed to TCDD and PAHs.
Environmental toxicology and pharmacology (2015-01-03)
Sonja Kaisarevic, Vanja Dakic, Jelena Hrubik, Branka Glisic, Urte Lübcke-von Varel, Kristina Pogrmic-Majkic, Svetlana Fa, Ivana Teodorovic, Werner Brack, Radmila Kovacevic
ABSTRACT
Rat hepatoma cells H4IIE were treated by 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) and polycyclic aromatic hydrocarbons (PAHs) (dibenz(a,h)anthracene, benzo(a)pyrene, benz(a)anthracene, chrysene), low-concentration mixtures of PAHs and TCDD, and environmental mixtures contaminated by PAHs and their derivatives. Expression of the gene battery comprising cytochrome P450 Cyp1a1, Cyp1a2, Cyp1b1, and glutathione-s-transferase Gsta2 and Gstp was investigated using quantitative real time polymerase chain reaction (qRT-PCR) analysis. The results revealed that TCDD induce Cyp1a1>Cyp1a2>Cyp1b1, while PAHs and PAH-containing environmental mixtures induce Cyp1a2>Cyp1a1>Cyp1b1 gene expression pattern. While low-concentration mixtures elicited a more pronounced response in comparison to single treatments, the typical gene expression patterns were not observed. In all samples, Gsta2 was predominantly expressed relative to Gstp. These findings indicate that differential Cyp1a1 and Cyp1a2 expression in the H4IIE cells might be used for detection of PAHs in highly contaminated environmental mixtures, but not in low-concentration mixtures of these compounds.
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Supelco
Benz[a]anthracene, analytical standard
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benz[a]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chrysene, analytical standard
Sigma-Aldrich
Fluorescamine, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Chrysene, BCR®, certified reference material
Benz[a]anthracene, BCR®, certified reference material
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Benz[a]anthracene, 99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Sigma-Aldrich
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)