MilliporeSigma
  • Myostatin and IGF-I signaling in end-stage human heart failure: a qRT-PCR study.
Myostatin and IGF-I signaling in end-stage human heart failure: a qRT-PCR study.
Journal of translational medicine (2015-01-17)
Júlia Aliz Baán, Zoltán V Varga, Przemyslaw Leszek, Mariusz Kuśmierczyk, Tamás Baranyai, László Dux, Péter Ferdinandy, Thomas Braun, Luca Mendler
ABSTRACT
Myostatin (Mstn) is a key regulator of heart metabolism and cardiomyocyte growth interacting tightly with insulin-like growth factor I (IGF-I) under physiological conditions. The pathological role of Mstn has also been suggested since Mstn protein was shown to be upregulated in the myocardium of end-stage heart failure. However, no data are available about the regulation of gene expression of Mstn and IGF-I in different regions of healthy or pathologic human hearts, although they both might play a crucial role in the pathomechanism of heart failure. In the present study, heart samples were collected from left ventricles, septum and right ventricles of control healthy individuals as well as from failing hearts of dilated (DCM) or ischemic cardiomyopathic (ICM) patients. A comprehensive qRT-PCR analysis of Mstn and IGF-I signaling was carried out by measuring expression of Mstn, its receptor Activin receptor IIB (ActRIIB), IGF-I, IGF-I receptor (IGF-IR), and the negative regulator of Mstn miR-208, respectively. Moreover, we combined the measured transcript levels and created complex parameters characterizing either Mstn- or IGF-I signaling in the different regions of healthy or failing hearts. We have found that in healthy control hearts, the ratio of Mstn/IGF-I signaling was significantly higher in the left ventricle/septum than in the right ventricle. Moreover, Mstn transcript levels were significantly upregulated in all heart regions of DCM but not ICM patients. However, the ratio of Mstn/IGF-I signaling remained increased in the left ventricle/septum compared to the right ventricle of DCM patients (similarly to the healthy hearts). In contrast, in ICM hearts significant transcript changes were detected mainly in IGF-I signaling. In parallel with these results miR-208 showed mild upregulation in the left ventricle of both DCM and ICM hearts. This is the first demonstration of a spatial asymmetry in the expression pattern of Mstn/IGF-I in healthy hearts, which is likely to play a role in the different growth regulation of left vs. right ventricle. Moreover, we identified Mstn as a massively regulated gene in DCM but not in ICM as part of possible compensatory mechanisms in the failing heart.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, Selectophore, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)