MilliporeSigma
  • Proinflammatory effect of trivalent arsenical species in a co-culture of Caco-2 cells and peripheral blood mononuclear cells.
Proinflammatory effect of trivalent arsenical species in a co-culture of Caco-2 cells and peripheral blood mononuclear cells.
Archives of toxicology (2014-05-28)
Marta Calatayud, José V Gimeno-Alcañiz, Vicenta Devesa, Dinoraz Vélez
ABSTRACT
Chronic exposure to inorganic arsenic (As) is associated with type 2 diabetes, cardiovascular diseases and cancer. Ingested inorganic As is transformed within the gastrointestinal tract and can give rise to more toxic species such as monomethylarsonous acid [MMA(III)] and dimethylarsinous acid [DMA(III)]. Thus, the intestinal epithelium comes into contact with toxic arsenical species, and the effects of such exposure upon epithelial function are not clear. The present study has evaluated the effect of 1 µM arsenite [As(III)], 0.1 µM MMA(III) and 1 µM DMA(III) upon the release of cytokines [interleukin-6 (IL6), IL8, tumor necrosis factor alpha (TNFα)], using a compartmentalized co-culture model with differentiated Caco-2 cells in the apical compartment and peripheral blood mononuclear cells in the basolateral compartment. In addition, the combined effect of arsenical species and lipopolysaccharide (LPS), both added into the apical compartment, has been analyzed. The results indicate that exposure to the arsenical forms induces a proinflammatory response. An increase in cytokine secretion into the basolateral compartment was observed, particularly as regards TNFα (up to 1,600 %). The cytokine levels on the apical side also increased, though to a lesser extent. As/LPS co-exposure significantly affected the proinflammatory response as compared to treatment with As alone. Treatment with DMA(III) and As/LPS co-exposure increased the permeability of the intestinal monolayer. In addition, As/LPS treatments enhanced As(III) and MMA(III) transport through the intestinal monolayer.
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 0.5 M in ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 0.02 M in ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 0.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylacetic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 0.1 M in isopropanol
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Pricing and availability is not currently available.
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Pricing and availability is not currently available.