MilliporeSigma
  • Discovery of novel Bcr-Abl inhibitors targeting myristoyl pocket and ATP site.
Discovery of novel Bcr-Abl inhibitors targeting myristoyl pocket and ATP site.
Bioorganic & medicinal chemistry (2014-12-04)
Jinyun Dong, Wen Lu, Xiaoyan Pan, Ping Su, Yaling Shi, Jinfeng Wang, Jie Zhang
ABSTRACT
Bcr-Abl plays an essential role in the pathogenesis and development of chronic myeloid leukaemia (CML). Inhibition of Bcr-Abl has great potential for therapeutic intervention in CML. In order to obtain novel and potent Bcr-Abl inhibitors, twenty seven 4,6-disubstituted pyrimidines were synthesized and evaluated herein. The biological results indicated that four compounds of them (C4, C5, C16, and C23) were potent Bcr-Abl inhibitors which were comparable to positive control. Moreover, C4 and C5 displayed promising antiproliferative activity against K562 cells. The results suggested that these 4,6-disubstituted pyrimidines could serve as promising leads for further optimization of Bcr-Abl inhibitors.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D