MilliporeSigma
  • Low-energy diets differing in fibre, red meat and coffee intake equally improve insulin sensitivity in type 2 diabetes: a randomised feasibility trial.
Low-energy diets differing in fibre, red meat and coffee intake equally improve insulin sensitivity in type 2 diabetes: a randomised feasibility trial.
Diabetologia (2014-11-27)
Bettina Nowotny, Lejla Zahiragic, Alessandra Bierwagen, Stefan Kabisch, Jan B Groener, Peter J Nowotny, Ann Kristin Fleitmann, Christian Herder, Giovanni Pacini, Iris Erlund, Rikard Landberg, Hans-Ulrich Haering, Andreas F H Pfeiffer, Peter P Nawroth, Michael Roden
ABSTRACT
Epidemiological studies have found that a diet high in fibre and coffee, but low in red meat, reduces the risk for type 2 diabetes. We tested the hypothesis that these nutritional modifications differentially improve whole-body insulin sensitivity (primary outcome) and secretion. Inclusion criteria were: age 18-69 years, BMI ≥ 30 kg/m(2), type 2 diabetes treated with diet, metformin or acarbose and known disease duration of ≤ 5 years. Exclusion criteria were: HbA1c >75 mmol/mol (9.0%), type 1 or secondary diabetes types and acute or chronic diseases including cancer. Patients taking any medication affecting the immune system or insulin sensitivity, other than metformin, were also excluded. Of 59 patients (randomised using randomisation blocks [four or six patients] with consecutive numbers), 37 (54% female) obese type 2 diabetic patients completed this controlled parallel-group 8-week low-energy dietary intervention. The participants consumed either a diet high in cereal fibre (whole grain wheat/rye: 30-50 g/day) and coffee (≥ 5 cups/day), and free of red meat (L-RISK, n = 17) or a diet low in fibre (≤ 10 g/day), coffee-free and high in red meat (≥ 150 g/day) diet (H-RISK, n = 20). Insulin sensitivity and secretion were assessed by hyperinsulinaemic-euglycaemic clamp and intravenous glucose tolerance tests with isotope dilution. Whole-body and organ fat contents were measured by magnetic resonance imaging and spectroscopy. Whole-body insulin sensitivity increased in both groups (mean [95% CI]) (H-RISK vs L-RISK: 0.8 [0.2, 1.4] vs 1.0 [0.4, 1.7]mg kg(-1) min(-1), p = 0.59), while body weight decreased (-4.8% [-6.1%, -3.5%] vs -4.6% [-6.0%, -3.3%], respectively). Hepatic insulin sensitivity remained unchanged, whereas hepatocellular lipid content fell in both groups (-7.0% [-9.6%, -4.5%] vs -6.7% [-9.5%, -3.9%]). Subcutaneous fat mass (-1,553 [-2,767, -340] cm(3) vs -751 [-2,047; 546] cm(3), respectively) visceral fat mass (-206 [-783, 371] cm(3) vs -241 [-856, 373] cm(3), respectively) and muscle fat content (-0.09% [-0.16%, -0.02%] vs -0.02% [-0.10%, 0.05%], respectively) decreased similarly. Insulin secretion remained unchanged, while the proinflammatory marker IL-18 decreased only after the L-RISK diet. No evidence of a difference between both low-energy diets was identified. Thus, energy restriction per se seems to be key for improving insulin action in phases of active weight loss in obese type 2 diabetic patients, with a potential improvement of subclinical inflammation with the L-RISK diet. Clinicaltrials.gov NCT01409330. This study was supported by the Ministry of Science and Research of the State of North Rhine-Westphalia (MIWF NRW), the German Federal Ministry of Health (BMG), the Federal Ministry for Research (BMBF) to the Center for Diabetes Research (DZD e.V.) and the Helmholtz Alliance Imaging and Curing Environmental Metabolic Diseases (ICEMED).
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, Plant-Derived, SyntheChol®