Merck
  • Losartan reduces myocardial interstitial fibrosis in diabetic cardiomyopathy rats by inhibiting JAK/STAT signaling pathway.
Losartan reduces myocardial interstitial fibrosis in diabetic cardiomyopathy rats by inhibiting JAK/STAT signaling pathway.
International journal of clinical and experimental pathology (2015-03-11)
Lijun Wang, Juan Li, Dajun Li
ABSTRACT
This study was designed to investigate the effect of losartan on the myocardial interstitial fibrosis in diabetic cardiomyopathy (DCM) rats. In this study, a total of 48 male Wister rats (3 groups of 16 animals each) were examined, including the control group, DCM group and losartan-treated (DCM + L) group. Control group was fed with standard diet (14 KJ/g); DCM group and losartan-treated (DCM + L) group were both fed with high glucose and fat diet (20 KJ/g). Diabetes was induced by streptozotocin (STZ) intraperitoneal injuction (IP, 30 mg/kg body weight). Rats of DCM + L group were treated with losartan (30 mg/kg body weight) daily by oral gavage for 16 weeks. Biochemical, hemodynamic, histological and western blotting analyses were performed. Compared with DCM rats, the quantity of p-JAK2 and p-STAT3 in myocardium of rats treated with losartan was lower, the expression of TGF-β1 was down-regulate, the content of collagen in myocardium decreased, LVSP and ± dp/dt increased, LVEDP decreased, the level of myocardial fibrosis reduced, and heart function improved evidently. Losartan has a protective effect on heart function against myocardial interstitial fibrosis of DCM by inhibiting JAK/STAT signaling pathway and lowering the expression of TGF-β1.
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, Selectophore, for ion-selective electrodes, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer