MilliporeSigma
  • Online background cleanup followed by high-performance liquid chromatography with tandem mass spectrometry for the analysis of perfluorinated compounds in human blood.
Online background cleanup followed by high-performance liquid chromatography with tandem mass spectrometry for the analysis of perfluorinated compounds in human blood.
Journal of separation science (2014-11-12)
Liangpo Liu, Jianwen She, Xueqin Zhang, Jie Zhang, Meiping Tian, Qingyu Huang, Syed Ali Musstjab Akber Shah Eqani, Heqing Shen
ABSTRACT
In this study, a novel method for the analysis of perfluorinated compounds in whole blood has been developed and validated. The method was developed by using a conventional reversed-phase C18 column as a trapping column for the elimination of background contamination and high-performance liquid chromatography with tandem mass spectrometry for the target compounds analysis. The trapping column provided fast online separation of the background contamination of perfluorinated compounds. In this developed method, the limits of detection for different perfluorinated compounds ranged from 0.06 to 0.14 ng/mL. It is notable that the limit of detection (0.07 ng/mL) for perfluorooctanoic acid was improved significantly after the elimination of background contamination. The method was also validated in terms of sensitivity, accuracy, and precision. The recoveries ranged from 66.8 to 111.9%, with relative standard deviations from 2.1 to 15.3%. Our preliminary data suggest that the novel method based on trapping column cleanup followed by high-performance liquid chromatography with tandem mass spectrometry could be applied in studies on the human exposure to perfluorinated compounds.
Product Description
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v)
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%