MilliporeSigma
  • Beta cell regeneration after single-round immunological destruction in a mouse model.
Beta cell regeneration after single-round immunological destruction in a mouse model.
Diabetologia (2014-10-24)
Jason M Tonne, Toshie Sakuma, Miguel Munoz-Gomez, Moustafa El Khatib, Michael A Barry, Yogish C Kudva, Yasuhiro Ikeda
ABSTRACT
Achieving a better understanding of beta cell regeneration after immunological destruction is crucial for the development of immunotherapy approaches for type 1 diabetes. In previous type 1 diabetes models, sustained immune activation eliminates regenerating beta cells, thus limiting the study of the regenerative capacity of beta cells upon immunological destruction. Here, we employed an adeno-associated virus 8 (AAV8) vector for beta cell-targeted overexpression of a foreign antigen to induce single-round immunological destruction of existing beta cells. Young and aged C57BL/6J mice were treated with AAV8 vectors expressing the foreign antigen luciferase. Islet inflammation and regeneration was observed at 3, 6, 10 and 22 weeks post-AAV delivery. In young C57BL/6J mice, robust humoral and cellular immune responses were developed towards antigen-expressing beta cells, leading to decreased beta cell mass. This was followed by beta cell mass replenishment, along with enhanced proliferation of insulin-positive cells, recruitment of nestin/CD34-positive endothelial cells, displacement of alpha cells and mobilisation of cytoplasmic neurogenin 3-positive cells. Mice with recovering beta cells showed normal or reduced fasting blood glucose levels and faster glucose clearance than controls. Although aged mice demonstrated similar responses to the treatment, they initially exhibited notable islet scarring and fluctuations in blood glucose levels, indicating that beta cell regeneration is slower in aged mice. Our hit-and-run, beta cell-targeted antigen expression system provides an opportunity to monitor the impact of single-round immunological beta cell destruction in animals with diverse genetic backgrounds or ageing status.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
In Stock
$195.00
50 kg
Estimated to ship on March 04, 2025
$6,017.00