MilliporeSigma
  • Preparation and characterization of anti-tissue factor single-chain variable fragment antibody for cancer diagnosis.
Preparation and characterization of anti-tissue factor single-chain variable fragment antibody for cancer diagnosis.
Cancer science (2014-10-15)
Ryuta Sato, Toshifumi Obonai, Ryo Tsumura, Kouhei Tsumoto, Yoshikatsu Koga, Masahiro Yasunaga, Yasuhiro Matsumura
ABSTRACT
Tissue factor (TF), which serves as the initiator of the extrinsic blood coagulation cascade, has been found to be overexpressed in various solid tumors, especially brain tumors, pancreatic cancer, and gastric cancer. Overexpression of TF is considered to contribute to the high incidence of thrombotic complications and poor prognosis in patients with such cancers. Therefore, detection or targeting of TF may be a promising approach for the diagnosis and treatment of solid tumors that are known to overexpress the protein. Here, we used the recombinant DNA technology to develop an anti-TF single-chain Fv (scFv) of small size and high affinity for its target. The biochemical characteristics of the anti-TF scFv were evaluated using surface plasmon resonance (SPR) sensing and flow cytometry. The data obtained showed that the affinity of the anti-TF scFv was 2.04 × 10(-8) (KD), and that the protein showed significant binding to the cancer cells. Then, Alexa 647-labeled anti-TF scFv and anti-TF IgG were administered to mice bearing chemically induced spontaneous tumors. The maximum tumor to background ratios of anti-TF scFv and anti-TF IgG were obtained 3 and 24 h after the injections, respectively. This study indicates anti-TF scFv may be suitable as an imaging probe for the diagnosis of solid tumors.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder