MilliporeSigma
  • Evolutionary Action Score of TP53 Identifies High-Risk Mutations Associated with Decreased Survival and Increased Distant Metastases in Head and Neck Cancer.
Evolutionary Action Score of TP53 Identifies High-Risk Mutations Associated with Decreased Survival and Increased Distant Metastases in Head and Neck Cancer.
Cancer research (2015-01-31)
David M Neskey, Abdullah A Osman, Thomas J Ow, Panagiotis Katsonis, Thomas McDonald, Stephanie C Hicks, Teng-Kuei Hsu, Curtis R Pickering, Alexandra Ward, Ameeta Patel, John S Yordy, Heath D Skinner, Uma Giri, Daisuke Sano, Michael D Story, Beth M Beadle, Adel K El-Naggar, Merrill S Kies, William N William, Carlos Caulin, Mitchell Frederick, Marek Kimmel, Jeffrey N Myers, Olivier Lichtarge
ABSTRACT
TP53 is the most frequently altered gene in head and neck squamous cell carcinoma, with mutations occurring in over two-thirds of cases, but the prognostic significance of these mutations remains elusive. In the current study, we evaluated a novel computational approach termed evolutionary action (EAp53) to stratify patients with tumors harboring TP53 mutations as high or low risk, and validated this system in both in vivo and in vitro models. Patients with high-risk TP53 mutations had the poorest survival outcomes and the shortest time to the development of distant metastases. Tumor cells expressing high-risk TP53 mutations were more invasive and tumorigenic and they exhibited a higher incidence of lung metastases. We also documented an association between the presence of high-risk mutations and decreased expression of TP53 target genes, highlighting key cellular pathways that are likely to be dysregulated by this subset of p53 mutations that confer particularly aggressive tumor behavior. Overall, our work validated EAp53 as a novel computational tool that may be useful in clinical prognosis of tumors harboring p53 mutations.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
Arginine, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®
Sign Into View Organizational & Contract Pricing