MilliporeSigma
  • Isolation and characterization of SSEA-4-positive subpopulation of human deciduous dental pulp cells.
Isolation and characterization of SSEA-4-positive subpopulation of human deciduous dental pulp cells.
Clinical oral investigations (2014-05-28)
Noriaki Kawanabe, Hiroaki Fukushima, Yoshihito Ishihara, Takeshi Yanagita, Hiroshi Kurosaka, Takashi Yamashiro
ABSTRACT
It is well accepted that stage-specific embryonic antigen (SSEA)-4 is an antigen that is useful to isolate adult stem cells analogous to embryonic stem cells. Therefore, in the present study, we investigated whether SSEA-4 can also be used as a marker to identify human deciduous dental pulp (D-DP) stem cells. Intact deciduous teeth were collected from healthy patients who were undergoing orthodontic treatment at Okayama University Hospital. Immunofluorescence analysis, flow cytometric analysis, and multilineage differentiation assay were performed to characterize SSEA-4+ D-DP cells. The D-DP cells had the characteristics of mesenchymal stem cells (MSCs), namely plastic adherence, specific surface antigen expression, and multipotent differentiation potential. SSEA-4 expression was detected in D-DP cells in vitro and ex vivo samples. A flow cytometric analysis demonstrated that 21.2 % of the D-DP cells were positive for SSEA-4. The SSEA-4+ clonal D-DP cells showed multilineage differentiation potential toward adipocytes, osteoblasts, and chondrocytes in vitro. In fact, 26.1 % (6/23) of the SSEA-4+ clonal D-DP cells showed adipogenic potential, 91.3 % (21/23) showed osteogenic potential, 91.3 % (21/23) showed chondrogenic potential, and 87.0 % (20/23) showed both osteogenic and chondrogenic potential. Thus, the majority of SSEA-4+ D-DP cells had the potential for multilineage differentiation. Hence, SSEA-4 appears to be a specific marker that can be used to identify D-DP stem cells. SSEA-4+ D-DP cells appear to be a promising source of stem cells for regenerative therapy.
Sigma-Aldrich
L-Proline, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, 99%
SAFC
L-Proline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%