MilliporeSigma
  • Association of genetic ancestry with breast cancer in ethnically diverse women from Chicago.
Association of genetic ancestry with breast cancer in ethnically diverse women from Chicago.
PloS one (2014-11-26)
Umaima Al-Alem, Garth Rauscher, Ebony Shah, Ken Batai, Abeer Mahmoud, Erin Beisner, Abigail Silva, Caryn Peterson, Rick Kittles
ABSTRACT
Non-Hispanic (nH) Black and Hispanic women are disproportionately affected by early onset disease, later stage, and with more aggressive, higher grade and ER/PR negative breast cancers. The purpose of this analysis was to examine whether genetic ancestry could account for these variation in breast cancer characteristics, once data were stratified by self-reported race/ethnicity and adjusted for potential confounding by social and behavioral factors. We used a panel of 100 ancestry informative markers (AIMs) to estimate individual genetic ancestry in 656 women from the "Breast Cancer Care in Chicago" study, a multi-ethnic cohort of breast cancer patients to examine the association between individual genetic ancestry and breast cancer characteristics. In addition we examined the association of individual AIMs and breast cancer to identify genes/regions that may potentially play a role in breast cancer disease disparities. As expected, nH Black and Hispanic patients were more likely than nH White patients to be diagnosed at later stages, with higher grade, and with ER/PR negative tumors. Higher European genetic ancestry was protective against later stage at diagnosis (OR 0.7 95%CI: 0.54-0.92) among Hispanic patients, and higher grade (OR 0.73, 95%CI: 0.56-0.95) among nH Black patients. After adjustment for multiple social and behavioral risk factors, the association with later stage remained, while the association with grade was not significant. We also found that the AIM SNP rs10954631 on chromosome 7 was associated with later stage (p = 0.02) and higher grade (p = 0.012) in nH Whites and later stage (p = 0.03) in nH Blacks. Non-European genetic ancestry was associated with later stage at diagnosis in ethnic minorities. The relation between genetic ancestry and stage at diagnosis may be due to genetic factors and/or unmeasured environmental factors that are overrepresented within certain racial/ethnic groups.
Product Description
Supelco
Water, for TOC analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Supelco
Water, suitable for ion chromatography
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Supelco
Water, ACS reagent, for ultratrace analysis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, HPLC Plus
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Sigma-Aldrich
Water, Deionized
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 μg
Estimated to ship on February 20, 2025
$444.00