MilliporeSigma
  • The HMGB1 protein sensitizes colon carcinoma cells to cell death triggered by pro-apoptotic agents.
The HMGB1 protein sensitizes colon carcinoma cells to cell death triggered by pro-apoptotic agents.
International journal of oncology (2014-12-02)
Chengcheng Christine Zhang, Georg Gdynia, Volker Ehemann, Wilfried Roth
ABSTRACT
The HMGB1 protein has multiple functions in tumor biology and can act both as a transcription factor and as a cytokine. HMGB1 is released during cell death, and in our previous studies we demonstrated that HMGB1 induces a distinct, necrosis-like cell death in glioblastoma. In epithelial malignant tumors such as colorectal cancer (CRC), the HMGB1-dependent effects show cross-talk with apoptotic signal transduction. Treatment of CRC cells with low concentrations of recombinant HMGB1 results in dose-dependent cytotoxicity which is morphologically characterized by the formation of giant mitochondria and does not share features of apoptosis. HMGB1-triggered cell death is associated with intracellular ROS release, and overexpression of Bcl-2 blocks both the increase of ROS as well as HMGB1-dependent cell death. Importantly, treatment with recombinant HMGB1 or overexpression of endogenous HMGB1 strongly sensitizes CRC cells to the cytotoxic activity of the pro-apoptotic death ligand TRAIL as well as the small molecule Bcl-2 family inhibitor ABT‑737. Moreover, treatment of CRC cells with TRAIL or ABT‑737 induces a release of endogenous HMGB1 into the extracellular space, and preincubation with glycyrrhizin, an HMGB1 inhibitor, significantly inhibits induction of cell death by TRAIL and ABT‑737, suggesting that HMGB1 functionally contributes to the execution of cell death triggered by pro-apoptotic agents. Finally, we investigated the expression of HMGB1 in human CRC tumor samples and found that loss of HMGB1 expression is associated with a more aggressive phenotype and a more advanced stage of disease in patients with CRC. Altogether, our findings demonstrate a functional link between cytotoxic signaling cascades triggered by HMGB1 and pro-apoptotic agents leading to an HMGB1-dependent sensitization to CRC cell death. Thus, a further evaluation of recombinant HMGB1 as part of an experimental combination treatment of CRC seems warranted.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)