MilliporeSigma
  • A 2-DE-based proteomic study on the toxicological effects of cisplatin in L02 cells.
A 2-DE-based proteomic study on the toxicological effects of cisplatin in L02 cells.
Environmental toxicology and pharmacology (2014-12-22)
Shu Liu, Wei Wang, Xueyi Zhou, Zongli Ding, Runhuan Gu
ABSTRACT
Cisplatin is a chemotherapeutic agent for the treatment of various cancers. In this study, cisplatin-induced effects were characterized in vitro model of human liver cells (L02) using 2-DE-based proteomics. Results indicated that different cisplatin treatments primarily induced disturbances in protein synthesis and oxidative stress via differential mechanisms. Since the experimental concentrations of cisplatin described a hormesis effect in cell proliferation of L02 cells, it was expected to reveal the hormesis effects using proteomic markers. However, only confilin-1 was commonly up-regulated in three concentrations of cisplatin treatments showing a hormesis effects with a U-shape regulation. These results were highly consistent with many other toxico-proteomic studies, indicating that the toxico-proteomic responses based on dose-dependent protein responses were incongruent with the theoretically linear or hormetic concentration-effect relationship. Our findings suggested that a macroscopic hormesis phenomenon on the cell proliferation could not be reflected by proteomic responses induced by cisplatin treatments.
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%